Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849306.5 | XP_854399.2 | 2 - 3 | 856 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |