Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119343.2 | XP_013974818.1 | 4 - 5 | 337 | 102 | |
XM_022403410.1 | XP_022259118.1 | 4 - 5 | 334 | 102 | ||
XM_022403411.1 | XP_022259119.1 | 4 - 5 | 316 | 102 | ||
XM_849901.5 | XP_854994.1 | 4 - 5 | 319 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |