Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022405650.1 | XP_022261358.1 | 1 - 2 | 246 | 58 | |
XM_022405651.1 | XP_022261359.1 | 1 - 2 | 246 | 58 | ||
XM_847733.4 | XP_852826.1 | 1 - 2 | 241 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |