Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287081.1 | NP_001274010.1 | 3 - 4 | 182 | 59 | |
GenBank mRNA | DN373580.1 | - | 3 - 4 | 182 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |