Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416373.1 | XP_022272081.1 | 4 - 5 | 455 | 69 | |
XM_022416374.1 | XP_022272082.1 | 3 - 4 | 304 | 69 | ||
XM_849112.5 | XP_854205.1 | 3 - 4 | 304 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Inventoried Probe spans exons |