Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416373.1 | XP_022272081.1 | 5 - 6 | 566 | 119 | |
XM_022416374.1 | XP_022272082.1 | 4 - 5 | 415 | 119 | ||
XM_849112.5 | XP_854205.1 | 4 - 5 | 415 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |