Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119530.2 | XP_013975005.1 | 3 - 4 | 693 | 85 | |
XM_022428135.1 | XP_022283843.1 | 3 - 4 | 690 | 85 | ||
XM_849268.5 | XP_854361.1 | 3 - 4 | 709 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |