Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638818.3 | XP_003638866.1 | 5 - 6 | 605 | 130 | |
XM_005616308.2 | XP_005616365.1 | 5 - 6 | 587 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |