Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414907.1 | XP_022270615.1 | 5 - 6 | 661 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |