Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850281.5 | XP_855374.2 | 2 - 3 | 251 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |