Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077588.2 | NP_001071056.2 | 3 - 4 | 377 | 55 | |
XM_005624362.3 | XP_005624419.1 | 3 - 4 | 438 | 55 | ||
GenBank mRNA | DN411459.1 | - | 2 - 3 | 242 | 55 | |
DN904672.1 | - | 3 - 4 | 379 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |