Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20H19orf77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849779.4 | XP_854872.2 | 3 - 4 | 325 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |