Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620856.3 | XP_005620913.1 | 4 - 5 | 647 | 81 | |
XM_014114114.2 | XP_013969589.1 | 3 - 4 | 884 | 81 | ||
XM_022419386.1 | XP_022275094.1 | 3 - 4 | 972 | 81 | ||
XM_848905.5 | XP_853998.2 | 3 - 4 | 517 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |