Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848130.4 | XP_853223.1 | 2 - 3 | 254 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |