Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637498.3 | XP_005637555.1 | 1 - 2 | 105 | 102 | |
XM_022411903.1 | XP_022267611.1 | 1 - 2 | 105 | 102 | ||
XM_851633.5 | XP_856726.2 | 1 - 2 | 105 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |