Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142670.1 | NP_001136142.1 | 3 - 4 | 391 | 74 | |
GenBank mRNA | AB467286.1 | - | 3 - 4 | 391 | 74 | |
AB467287.1 | - | 3 - 4 | 321 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |