Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252264.1 | NP_001239193.1 | 1 | 394 | 69 | |
NM_001252265.1 | NP_001239194.1 | 2 | 373 | 69 | ||
GenBank mRNA | DN370387.1 | - | 2 | 373 | 69 | |
DN371621.1 | - | 349 | 69 | |||
DN865199.1 | - | 1 | 108 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |