Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130437.2 | NP_001123909.2 | 3 - 4 | 544 | 65 | |
XM_005623168.3 | XP_005623225.1 | 3 - 4 | 520 | 65 | ||
GenBank mRNA | DN380583.1 | - | 3 - 4 | 428 | 65 | |
DN400224.1 | - | 3 - 4 | 533 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |