Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_532457.6 | XP_532457.3 | 1 - 2 | 338 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |