Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_844580.4 | XP_849673.1 | 418 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |