Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619537.3 | XP_005619594.2 | 115 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |