Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEPCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419643.1 | XP_022275351.1 | 1661 | 87 | ||
XM_546957.6 | XP_546957.2 | 2082 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris |