Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621082.3 | XP_005621139.1 | 964 | 130 | ||
XM_005621084.3 | XP_005621141.1 | 887 | 130 | |||
XM_022419713.1 | XP_022275421.1 | 969 | 130 | |||
XM_022419714.1 | XP_022275422.1 | 752 | 130 | |||
XM_855762.5 | XP_860855.1 | 880 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |