Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115777.2 | XP_013971252.1 | 1 - 2 | 304 | 78 | |
XM_854240.5 | XP_859333.2 | 1 - 2 | 300 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |