Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313777.1 | NP_001300706.1 | 4 - 5 | 381 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |