Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632147.3 | XP_005632204.1 | 1281 | 80 | ||
XM_005632148.3 | XP_005632205.1 | 963 | 80 | |||
XM_005632150.3 | XP_005632207.1 | 452 | 80 | |||
XM_014121565.2 | XP_013977040.1 | 633 | 80 | |||
XM_022406363.1 | XP_022262071.1 | 662 | 80 | |||
XM_844730.5 | XP_849823.2 | 636 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |