Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534781.6 | XP_534781.2 | 6 - 7 | 743 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |