Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_539141.5 | XP_539141.2 | 313 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |