Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416467.1 | XP_022272175.1 | 2 - 3 | 659 | 152 | |
XM_549183.6 | XP_549183.4 | 3 - 4 | 768 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |