Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641441.3 | XP_005641498.1 | 2 | 605 | 78 | |
XM_005641442.3 | XP_005641499.1 | 2 | 456 | 78 | ||
XM_014111872.2 | XP_013967347.1 | 2 | 768 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |