Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_542952.6 | XP_542952.3 | 8 - 9 | 1024 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |