Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639779.2 | XP_003639827.1 | 18 - 19 | 2498 | 115 | |
XM_003639780.3 | XP_003639828.1 | 18 - 19 | 2369 | 115 | ||
XM_005632788.3 | XP_005632845.1 | 18 - 19 | 2344 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |