Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGGBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638739.3 | XP_005638796.1 | 171 | 148 | ||
XM_535549.7 | XP_535549.1 | 169 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |