Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_543942.5 | XP_543942.2 | 1 - 2 | 158 | 134 | |
XM_844638.4 | XP_849731.1 | 1 - 2 | 158 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |