Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110793.2 | XP_013966268.1 | 4 | 2180 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |