Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120729.2 | XP_013976204.1 | 3 - 4 | 625 | 86 | |
XM_014120730.2 | XP_013976205.1 | 3 - 4 | 515 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |