Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848250.5 | XP_853343.1 | 2 | 399 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |