Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002940.1 | NP_001002940.1 | 4 | 517 | 148 | |
XM_005620606.3 | XP_005620663.1 | 4 | 1745 | 148 | ||
GenBank mRNA | AY217754.1 | - | 4 | 517 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Amplicon spans exons and probe does not span exons |