Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630713.3 | XP_005630770.1 | 11 - 12 | 1185 | 147 | |
XM_022404930.1 | XP_022260638.1 | 10 - 11 | 1094 | 147 | ||
XM_022404931.1 | XP_022260639.1 | 10 - 11 | 1074 | 147 | ||
XM_022404932.1 | XP_022260640.1 | 10 - 11 | 1081 | 147 | ||
XM_844483.5 | XP_849576.1 | 11 - 12 | 1167 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Recommended Assay Canis familiaris Probe spans exons |