Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_540511.4 | XP_540511.2 | 2 | 235 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |