Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204407.1 | NP_001191336.1 | 2 | 443 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Amplicon spans exons and probe does not span exons |