Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536465.6 | XP_536465.3 | 3 | 3108 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |