Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546152.5 | XP_546152.2 | 6 | 1364 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |