Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195843.1 | NP_001182772.1 | 1 | 553 | 108 | |
XM_005635607.3 | XP_005635664.1 | 3 | 1610 | 108 | ||
XM_022409542.1 | XP_022265250.1 | 2 | 1860 | 108 | ||
XM_022409544.1 | XP_022265252.1 | 2 | 1864 | 108 | ||
XM_022409545.1 | XP_022265253.1 | 2 | 1862 | 108 | ||
XM_022409546.1 | XP_022265254.1 | 2 | 1871 | 108 | ||
GenBank mRNA | AF068283.1 | - | 1 | 554 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |