Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548063.5 | XP_548063.2 | 684 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |