Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544656.6 | XP_544656.2 | 2 - 3 | 261 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |