Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_544656.6 | XP_544656.2 | 4 - 5 | 404 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |