Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619640.3 | XP_005619697.1 | 4533 | 193 | ||
XM_005619642.3 | XP_005619699.1 | 1761 | 193 | |||
XM_014113349.2 | XP_013968824.1 | 2984 | 193 | |||
XM_022418166.1 | XP_022273874.1 | 1936 | 193 | |||
XM_022418167.1 | XP_022273875.1 | 1997 | 193 | |||
XM_022418168.1 | XP_022273876.1 | 1839 | 193 | |||
XM_546469.6 | XP_546469.2 | 1636 | 193 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris |