Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427957.1 | XP_022283665.1 | 2 - 3 | 238 | 145 | |
XM_847756.6 | XP_852849.3 | 2 - 3 | 348 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |