Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313829.1 | NP_001300758.1 | 3 | 416 | 93 | |
XM_005631378.3 | XP_005631435.1 | 3 | 695 | 93 | ||
XM_005631379.3 | XP_005631436.1 | 2 | 277 | 93 | ||
XM_022405076.1 | XP_022260784.1 | 3 | 273 | 93 | ||
XM_022405077.1 | XP_022260785.1 | 3 | 510 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |